Learn More
Medchemexpress LLC HY-10256A 10mg Medchemexpress, SB 203580 (hydrochloride) CAS:869185-85-3 Purity:>98%
SDP
Catalog No. 501967992
Encompass_Preferred
Medchemexpress, HY-10256A 10mg SB 203580 (HCl) CAS:869185-85-3 SB 203580 hydrochloride (RWJ 64809 hydrochloride) is a widely used p38 MAPK inhibitor. SB 203580 hydrochloride (RWJ 64809 hydrochloride) inhibits SAPK2a/p38 and SAPK2b/p38β2, with IC50s of 50 nM and 500 nM, respectively. LCK, GSK3β and PKBα are also inhibited by SB 203580 hydrochloride (RWJ 64809 hydrochloride), but the IC50s are 100-500-fold higher than that for SAPK2a/p38. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7992 Supplier Medchemexpress LLC Supplier No. HY10256A10MG
Medchemexpress, HY-10256A 10mg SB 203580 (HCl) CAS:869185-85-3 SB 203580 hydrochloride (RWJ 64809 hydrochloride) is a widely used p38 MAPK inhibitor. SB 203580 hydrochloride (RWJ 64809 hydrochloride) inhibits SAPK2a/p38 and SAPK2b/p38β2, with IC50s of 50 nM and 500 nM, respectively. LCK, GSK3β and PKBα are also inhibited by SB 203580 hydrochloride (RWJ 64809 hydrochloride), but the IC50s are 100-500-fold higher than that for SAPK2a/p38. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.