Learn More
Cayman Chemical SB 203580 (hydrochloride), Purity: ≥95%, CAS: 869185-85-3, 50 mg
SDP
Catalog No. 502953509
Encompass_Preferred
A formulation with greater solubility in organic solvents than standard SB 203580
Catalog No. 50-295-3509 Supplier Cayman Chemical Supplier No. 1334450 MG
A formulation with greater solubility in organic solvents than standard SB 203580