Learn More
Abcam SB 204741, 5-HT2B receptor antagonist, 10MG
Catalog No. 502707446
Encompass_Preferred
MW 286.35 g/mol, Purity >99%. Selective 5-HT₂B receptor antagonist. High affinity for 5-HT₂B receptors (pKi = 7.85), some activity at 5-HT₂C receptors (pKi = 5.8).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7446 Supplier Abcam Supplier No. AB12059510MG
MW 286.35 g/mol, Purity >99%. Selective 5-HT₂B receptor antagonist. High affinity for 5-HT₂B receptors (pKi = 7.85), some activity at 5-HT₂C receptors (pKi = 5.8).
The product is subject to the following: Abcam Restricted Use Statement