Learn More
Medchemexpress LLC SB 204990 | 154566-12-8 | 99.8% | 389.27 | 100 MG
SDP
Catalog No. 5000394080
Encompass_Preferred
SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme. It is for research use only. SB 204990 treatment leads to a decrease in cytosolic Ac-CoA levels, which is expected to decrease acetylated and active β-catenin levels. When administered orally to rats, it is absorbed into the systemic circulation, causing a dose-related decrease in plasma cholesterol and triglyceride levels. In dogs, it also decreases plasma cholesterol and triglyceride levels, preferentially decreasing low-density lipoprotein compared with high-density lipoprotein cholesterol levels.
  • Potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.
  • Decreases cytosolic Ac-CoA levels.
  • Reduces acetylated and active β-catenin levels.
  • Decreases plasma cholesterol and triglyceride levels in rats and dogs.
  • Preferentially decreases low-density lipoprotein compared with high-density lipoprotein cholesterol levels in dogs.
Catalog No. 50-003-94080 Supplier Medchemexpress LLC Supplier No. HY16450100MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme. It is for research use only. SB 204990 treatment leads to a decrease in cytosolic Ac-CoA levels, which is expected to decrease acetylated and active β-catenin levels. When administered orally to rats, it is absorbed into the systemic circulation, causing a dose-related decrease in plasma cholesterol and triglyceride levels. In dogs, it also decreases plasma cholesterol and triglyceride levels, preferentially decreasing low-density lipoprotein compared with high-density lipoprotein cholesterol levels.
  • Potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.
  • Decreases cytosolic Ac-CoA levels.
  • Reduces acetylated and active β-catenin levels.
  • Decreases plasma cholesterol and triglyceride levels in rats and dogs.
  • Preferentially decreases low-density lipoprotein compared with high-density lipoprotein cholesterol levels in dogs.