Learn More
Medchemexpress LLC SB 206553 hydrochloride | 1197334-04-5 | 99.4% | 328.80 g/mol | C17H17ClN4O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10313525MG
Encompass_Preferred
SB 206553 hydrochloride is a research compound that selectively antagonizes 5-HT2B and 5-HT2C serotonin receptors and is used in pharmacology and neuroscience studies to probe serotonergic signaling.
  • Selective 5-HT2B/5-HT2C receptor antagonist/inverse agonist
  • High purity suitable for research applications
  • Solid, light yellow to yellow appearance for easy handling
  • Stable when stored sealed and protected from moisture at -20°C
  • Available in small milligram quantities for preclinical studies
Catalog No. 50-002-45789