Learn More
Abcam SB 206553 hydrochloride, 5-HT2B and 5-HT2C receptor antagonist, 50MG
Catalog No. 502707492
Encompass_Preferred
MW 292.33 Da, Purity >99%. Potent and selective 5-HT2B and 5-HT2C serotonin receptor antagonist (pKi values are 7.92 (5-HT2C), 5.78 (5-HT2A), 7.65 (5-HT2B), 1A), 1D), 1E) and 1F). Centrally active following oral administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7492 Supplier Abcam Supplier No. AB12055650MG
MW 292.33 Da, Purity >99%. Potent and selective 5-HT2B and 5-HT2C serotonin receptor antagonist (pKi values are 7.92 (5-HT2C), 5.78 (5-HT2A), 7.65 (5-HT2B), 1A), 1D), 1E) and 1F). Centrally active following oral administration.
The product is subject to the following: Abcam Restricted Use Statement