Learn More
TARGETMOL CHEMICALS INC SB-209670 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T345415MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-209670 is a selective endothelin receptor antagonist that inhibits the activity of ET(B) receptors in rat vascular endothelium and vascular smooth muscle. purity: 99%
Catalog No. 50-258-0257