Learn More
TARGETMOL CHEMICALS INC SB-216 5MG
SDP
Catalog No. 502579589
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-216 is able to be used for the research of cancer which is a potent inhibitor of tubulin polymerization. SB-216 exhibits strong antiproliferative potency in a panel of human cancer cell lines including melanoma breast cancer and lung cancer [1]. purity: 100%
Catalog No. 50-257-9589 Supplier TARGETMOL CHEMICALS INC Supplier No. T607575MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-216 is able to be used for the research of cancer which is a potent inhibitor of tubulin polymerization. SB-216 exhibits strong antiproliferative potency in a panel of human cancer cell lines including melanoma breast cancer and lung cancer [1]. purity: 100%