Learn More
Abcam SB 216641 hydrochloride, h5-HT1B antagonist, 50MG
Catalog No. 502707461
Encompass_Preferred
MW 523.03 g/mol, Purity >98%. Selective h5-HT₁B receptor antagonist. 25-fold selectivity over h5-HT₁D. Little or no affinity for a range of other receptor subtypes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7461 Supplier Abcam Supplier No. AB12058350MG
MW 523.03 g/mol, Purity >98%. Selective h5-HT₁B receptor antagonist. 25-fold selectivity over h5-HT₁D. Little or no affinity for a range of other receptor subtypes.
The product is subject to the following: Abcam Restricted Use Statement