Learn More
Cayman Chemical SB-216641 (hydrochloride), Purity: ≥98%, CAS: 193611-67-5, 10 mg
SDP
Catalog No. 502950121
Encompass_Preferred
A 5-HT1B receptor antagonist; binds to 5-HT1B receptors (Ki = 1 nM); selective for 5-HT1B over 5-HT1A, 5-HT1D, 5-HT1E, and 5-HT1F receptors in radioligand binding assays (Kis = 500, 25, >1,000, and >1,000 nM, respectively); reduces 5-HT-induced inhibition of forskolin-induced cAMP production in CHO cells expressing human 5-HT1B receptors from 1-1,000 nM
Catalog No. 50-295-0121 Supplier Cayman Chemical Supplier No. 3283710 MG
A 5-HT1B receptor antagonist; binds to 5-HT1B receptors (Ki = 1 nM); selective for 5-HT1B over 5-HT1A, 5-HT1D, 5-HT1E, and 5-HT1F receptors in radioligand binding assays (Kis = 500, 25, >1,000, and >1,000 nM, respectively); reduces 5-HT-induced inhibition of forskolin-induced cAMP production in CHO cells expressing human 5-HT1B receptors from 1-1,000 nM