Learn More
Selleck Chemical LLC SB216763 5mg 280744-09-4
SDP
Catalog No. 508408
Encompass_Preferred
SB216763 is a potent and selective GSK-3 inhibitor with IC50 of 34.3 nM for GSK-3? and equally effective at inhibiting human GSK-3?. SB216763 activates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-840-8 Supplier Selleck Chemical LLC Supplier No. S10755MG
SB216763 is a potent and selective GSK-3 inhibitor with IC50 of 34.3 nM for GSK-3? and equally effective at inhibiting human GSK-3?. SB216763 activates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.