Learn More
Abcam SB 216763, GSK3 inhibitor, 50MG
Catalog No. 502707792
Encompass_Preferred
MW 371.2 Da, Purity >98%. Potent and selective GSK3 inhibitor (Ki= 9 nM for GSK-3α). Has little activity against 24 other protein kinases (IC₅₀ > 10 μM). Stimulates glycogen synthesis, gene transcription and is cardio- and neuroprotective.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7792 Supplier Abcam Supplier No. AB12020250MG
MW 371.2 Da, Purity >98%. Potent and selective GSK3 inhibitor (Ki= 9 nM for GSK-3α). Has little activity against 24 other protein kinases (IC₅₀ > 10 μM). Stimulates glycogen synthesis, gene transcription and is cardio- and neuroprotective.
The product is subject to the following: Abcam Restricted Use Statement