Learn More
Abcam SB 216763, GSK3 inhibitor, 1MG
Catalog No. 502707793
Encompass_Preferred
MW 371.2 Da, Purity >98%. Potent and selective GSK3 inhibitor (Ki= 9 nM for GSK-3α). Has little activity against 24 other protein kinases (IC₅₀ > 10 μM). Stimulates glycogen synthesis, gene transcription and is cardio- and neuroprotective.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7793 Supplier Abcam Supplier No. AB1202021MG
MW 371.2 Da, Purity >98%. Potent and selective GSK3 inhibitor (Ki= 9 nM for GSK-3α). Has little activity against 24 other protein kinases (IC₅₀ > 10 μM). Stimulates glycogen synthesis, gene transcription and is cardio- and neuroprotective.
The product is subject to the following: Abcam Restricted Use Statement