Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-224289 (hydrochloride) | 180084-26-8 | 99.0% | 557.08 g/mol | C32H33ClN4O3 | 100 MG
SDP

Catalog No. 5000211839
Encompass_Preferred

SB-224289 hydrochloride is a research-grade selective 5-HT1B receptor antagonist reported to have anxiolytic effects. Supplied as the hydrochloride salt in solid form, it is intended for laboratory research use only and is used in pharmacology and neuroscience studies of 5-HT1B receptor function.

  • High purity: 99.0%.
  • Cas number: 180084-26-8.
  • Molecular formula C32H33ClN4O3; molecular weight 557.08 g/mol.
  • Supplied as a 100 MG solid pack.
  • Recommended storage under nitrogen at -20°C; in solution -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-11839 Supplier Medchemexpress LLC Supplier No. HY101105A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB-224289 hydrochloride is a research-grade selective 5-HT1B receptor antagonist reported to have anxiolytic effects. Supplied as the hydrochloride salt in solid form, it is intended for laboratory research use only and is used in pharmacology and neuroscience studies of 5-HT1B receptor function.

  • High purity: 99.0%.
  • Cas number: 180084-26-8.
  • Molecular formula C32H33ClN4O3; molecular weight 557.08 g/mol.
  • Supplied as a 100 MG solid pack.
  • Recommended storage under nitrogen at -20°C; in solution -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.