Learn More
Medchemexpress LLC SB-224289 (hydrochloride) | 180084-26-8 | MFCD03788209 | 99.0% | 557.08 | C32H33ClN4O3 | 50 MG
Supplier: Medchemexpress LLC HY101105A50MG
SB-224289 hydrochloride is a research-grade hydrochloride salt that acts as a selective 5-HT1B (serotonin 1B) receptor antagonist, used in preclinical studies to probe serotonergic signaling and related behavioral effects such as anxiolysis. The compound is supplied with high stated purity and should be stored under low-temperature, moisture-free conditions.
- Selective 5-HT1B receptor antagonist for targeted pharmacological studies.
- Useful for probing serotonergic signaling and anxiolytic-related behavior in preclinical research.
- High stated purity (99.0%).
- Supplied as the hydrochloride salt in solid form for stability.
- Recommended storage at low temperature, protected from moisture.
- Available in small research quantities suitable for laboratory experiments.