Learn More
Cayman Chemical SB-224289, Purity: ≥98%, CAS: 180083-23-2, 50 mg
SDP
Catalog No. 502967038
Encompass_Preferred
A potent 5-HT1B inverse agonist (pKi = 8.16) that displays >75-fold selectivity for human 5-HT1B over all other 5-HT receptors
Catalog No. 50-296-7038 Supplier Cayman Chemical Supplier No. 1703650 MG
A potent 5-HT1B inverse agonist (pKi = 8.16) that displays >75-fold selectivity for human 5-HT1B over all other 5-HT receptors