Learn More
Apexbio Technology LLC SB 225002, 10mg. Cas: 182498-32-4 MFCD: MFCD00954637. CXCR2 antagonist, potent and selective
SDP
Supplier:  Apexbio Technology LLC B820010
Encompass_Preferred
SB 225002 is a potent and selective CXCR2 chemokine receptor antagonist with IC50 value of 22nM [1].CXCR2 is a member of the G-protein-coupled receptor family, which is responsible for neutrophil chemotaxis and margination induced by IL-8 [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5494