Learn More
Medchemexpress LLC SB 239063 | 193551-21-2 | >99.2% | 368.40 | 1 ML
SDP
Catalog No. 5000343740
Encompass_Preferred
SB 239063 is a potent, selective, and orally active p38 MAPK inhibitor. It exhibits an IC50 of 44 nM for recombinant purified human p38α and has equipotent inhibitory activity against p38α and p38β. This compound does not affect p38γ or p38δ. It demonstrates anti-asthma activity and can be utilized to enhance memory impaired due to aging or medical conditions like Alzheimer's Disease (AD), as it can penetrate the blood-brain barrier (BBB).
  • Potent, selective, and orally active p38 MAPK inhibitor.
  • Exhibits equipotent inhibitory activity against p38α and p38β.
  • Demonstrates anti-asthma activity.
  • Can enhance memory.
  • Penetrates the blood-brain barrier (BBB).
Catalog No. 50-003-43740 Supplier Medchemexpress LLC Supplier No. HY110681MG
SB 239063 is a potent, selective, and orally active p38 MAPK inhibitor. It exhibits an IC50 of 44 nM for recombinant purified human p38α and has equipotent inhibitory activity against p38α and p38β. This compound does not affect p38γ or p38δ. It demonstrates anti-asthma activity and can be utilized to enhance memory impaired due to aging or medical conditions like Alzheimer's Disease (AD), as it can penetrate the blood-brain barrier (BBB).
  • Potent, selective, and orally active p38 MAPK inhibitor.
  • Exhibits equipotent inhibitory activity against p38α and p38β.
  • Demonstrates anti-asthma activity.
  • Can enhance memory.
  • Penetrates the blood-brain barrier (BBB).