Learn More
TARGETMOL CHEMICALS INC SB 242084 10MG
SDP
Catalog No. 502576333
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB 242084 dihydrochloride hydrate is a psychoactive drug and research chemical which acts as a selective antagonist for the 5HT2C receptor. purity: 99%
Catalog No. 50-257-6333 Supplier TARGETMOL CHEMICALS INC Supplier No. T727710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB 242084 dihydrochloride hydrate is a psychoactive drug and research chemical which acts as a selective antagonist for the 5HT2C receptor. purity: 99%