Learn More
Medchemexpress LLC SB 242084 dihydrochloride | 1049747-87-6 | MFCD02684417 | 98.1% | C21H21Cl3N4O2 | 10MG
SDP
Catalog No. 5000261927
Encompass_Preferred
SB 242084 dihydrochloride is a selective, high-affinity 5-HT2C receptor antagonist used in neuroscience research. It crosses the blood-brain barrier and is used to study serotonergic regulation of dopaminergic neurons, behavioral effects, and metabolic changes. Provided as a dihydrochloride salt and available in solution formats, it is intended for research use only.
  • Selective 5-HT2C receptor antagonist
  • High affinity (pKi = 9.0)
  • Crosses the blood-brain barrier
  • Suitable for in vitro and in vivo studies
  • Available as solid salt and DMSO solution formats
  • Intended for research use only
Catalog No. 50-002-61927 Supplier Medchemexpress LLC Supplier No. HY13409A10MG
SB 242084 dihydrochloride is a selective, high-affinity 5-HT2C receptor antagonist used in neuroscience research. It crosses the blood-brain barrier and is used to study serotonergic regulation of dopaminergic neurons, behavioral effects, and metabolic changes. Provided as a dihydrochloride salt and available in solution formats, it is intended for research use only.
  • Selective 5-HT2C receptor antagonist
  • High affinity (pKi = 9.0)
  • Crosses the blood-brain barrier
  • Suitable for in vitro and in vivo studies
  • Available as solid salt and DMSO solution formats
  • Intended for research use only