Learn More
Medchemexpress LLC HY-18306 5mg Medchemexpress, SB 242235 CAS:193746-75-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY183065MG
Encompass_Preferred
Medchemexpress, HY-18306 5mg SB 242235 CAS:193746-75-7 SB-242235 is a potent and selective p38 MAP kinase inhibitor, with an IC50 of 1.0 μM in primary human chondrocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2035