Learn More
Abcam SB 243213 dihydrochloride, 5HT2C receptor antagonist, 10MG
Catalog No. 502706712
Encompass_Preferred
MW 464.9 Da, Purity >99%. Potent, selective 5-HT2C receptor antagonist (Ki = 1 nM). Neurological agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6712 Supplier Abcam Supplier No. AB14129110MG
MW 464.9 Da, Purity >99%. Potent, selective 5-HT2C receptor antagonist (Ki = 1 nM). Neurological agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement