Learn More
Medchemexpress LLC SB-258585 hydrochloride | 1216468-02-8 | 99.7% | 523.82 g/mol | C18H23ClIN3O3S | 10 MG
SDP
Catalog No. 5000246107
Encompass_Preferred
SB-258585 hydrochloride is a research compound used as a high-affinity ligand and antagonist for the serotonin 5-HT6 receptor (CAS 1216468-02-8). Supplied as the hydrochloride salt, it has molecular formula C18H23ClIN3O3S, molecular weight 523.82 g/mol, and is provided for in vitro and ex vivo pharmacological and receptor-binding studies.
  • High-affinity antagonist of the serotonin 5-HT6 receptor.
  • High reported purity, suitable for analytical and biological assays.
  • Appropriate for radioligand binding and receptor labeling studies.
  • Supplied in small-mass research quantities for laboratory use.
  • Molecular weight 523.82 g/mol and defined molecular formula.
Catalog No. 50-002-46107 Supplier Medchemexpress LLC Supplier No. HY10314110MG
SB-258585 hydrochloride is a research compound used as a high-affinity ligand and antagonist for the serotonin 5-HT6 receptor (CAS 1216468-02-8). Supplied as the hydrochloride salt, it has molecular formula C18H23ClIN3O3S, molecular weight 523.82 g/mol, and is provided for in vitro and ex vivo pharmacological and receptor-binding studies.
  • High-affinity antagonist of the serotonin 5-HT6 receptor.
  • High reported purity, suitable for analytical and biological assays.
  • Appropriate for radioligand binding and receptor labeling studies.
  • Supplied in small-mass research quantities for laboratory use.
  • Molecular weight 523.82 g/mol and defined molecular formula.