Learn More
Medchemexpress LLC SB-258585 hydrochloride | 1216468-02-8 | 99.7% | 523.82 g/mol | C18H23ClIN3O3S | 25 MG
SDP
Catalog No. 5000247806
Encompass_Preferred
SB-258585 hydrochloride is a research-grade small molecule that binds with high affinity to the 5-HT6 (serotonin 6) receptor and is commonly used in receptor labeling and pharmacological studies.
  • High affinity for the 5-HT6 receptor.
  • Useful for labeling recombinant and native 5-HT6 receptors.
  • High reported purity (about 99.7%).
  • Provided as the hydrochloride salt for improved stability.
  • Available in small research pack sizes, suitable for laboratory assays.
Catalog No. 50-002-47806 Supplier Medchemexpress LLC Supplier No. HY10314125MG
SB-258585 hydrochloride is a research-grade small molecule that binds with high affinity to the 5-HT6 (serotonin 6) receptor and is commonly used in receptor labeling and pharmacological studies.
  • High affinity for the 5-HT6 receptor.
  • Useful for labeling recombinant and native 5-HT6 receptors.
  • High reported purity (about 99.7%).
  • Provided as the hydrochloride salt for improved stability.
  • Available in small research pack sizes, suitable for laboratory assays.