Learn More
Medchemexpress LLC Sb-258585 hydrochloride | 1216468-02-8 | 99.7% | 523.82 g·mol⁻¹ | C18H23ClIN3O3S | 5 MG
SDP
Catalog No. 5000246135
Encompass_Preferred
SB-258585 hydrochloride is the hydrochloride salt of a selective, high-affinity antagonist for the serotonin 5-HT6 receptor. Supplied as a research-grade powder, it is intended for in vitro receptor binding and pharmacology studies and is characterized by high purity and defined storage conditions.
  • Selective 5-HT6 receptor antagonist with high affinity.
  • Suitable for in vitro receptor binding and pharmacological assays.
  • Provided as a hydrochloride salt in powder form for accurate dosing.
  • High purity (>99.7%) for reproducible results.
  • Stable when stored under recommended conditions (powder: -20°C).
Catalog No. 50-002-46135 Supplier Medchemexpress LLC Supplier No. HY1031415MG
SB-258585 hydrochloride is the hydrochloride salt of a selective, high-affinity antagonist for the serotonin 5-HT6 receptor. Supplied as a research-grade powder, it is intended for in vitro receptor binding and pharmacology studies and is characterized by high purity and defined storage conditions.
  • Selective 5-HT6 receptor antagonist with high affinity.
  • Suitable for in vitro receptor binding and pharmacological assays.
  • Provided as a hydrochloride salt in powder form for accurate dosing.
  • High purity (>99.7%) for reproducible results.
  • Stable when stored under recommended conditions (powder: -20°C).