Learn More
Cayman Chemical SB-258585hydrochlorIde 10mg
SDP
Catalog No. 502966493
Encompass_Preferred
A potent, selective antagonist of the serotonin 5-HT6 receptor (pKi = 8.53); has been used in cells and in animals
Catalog No. 50-296-6493 Supplier Cayman Chemical Supplier No. 1741610 MG
A potent, selective antagonist of the serotonin 5-HT6 receptor (pKi = 8.53); has been used in cells and in animals