Learn More
Cayman Chemical SB-258585 (hydrochloride), Purity: ≥95%, 50 mg
SDP
Catalog No. 502967135
Encompass_Preferred
A potent, selective antagonist of the serotonin 5-HT6 receptor (pKi = 8.53); has been used in cells and in animals
Catalog No. 50-296-7135 Supplier Cayman Chemical Supplier No. 1741650 MG
A potent, selective antagonist of the serotonin 5-HT6 receptor (pKi = 8.53); has been used in cells and in animals