Learn More
TARGETMOL CHEMICALS INC SB 258719 10MG
SDP
Catalog No. 502577408
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB 258719 is a selective antagonist of 5-HT7 receptor with pKi of 7.5. purity: 98%
Catalog No. 50-257-7408 Supplier TARGETMOL CHEMICALS INC Supplier No. T1285510MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB 258719 is a selective antagonist of 5-HT7 receptor with pKi of 7.5. purity: 98%