Learn More
Medchemexpress LLC SB 258719 | 195199-95-2 | 338.51 | 100 MG
SDP
Catalog No. 5000448801
Encompass_Preferred
SB 258719 is a selective 5-HT7 receptor antagonist with a high affinity (pKi=7.5) for the receptor. It is used in the research of cancer and neurological diseases. For research use only; not sold to patients.
  • Selective 5-HT7 receptor antagonist with high affinity (pKi=7.5)
  • Used for research of cancer and neurological disease
  • Produces a concentration-related rightward-shift of the 5-CT concentration-response curve
  • Significantly attenuates 5-CT-induced hypothermia in mice
Catalog No. 50-004-48801 Supplier Medchemexpress LLC Supplier No. HYU00443100MG
SB 258719 is a selective 5-HT7 receptor antagonist with a high affinity (pKi=7.5) for the receptor. It is used in the research of cancer and neurological diseases. For research use only; not sold to patients.
  • Selective 5-HT7 receptor antagonist with high affinity (pKi=7.5)
  • Used for research of cancer and neurological disease
  • Produces a concentration-related rightward-shift of the 5-CT concentration-response curve
  • Significantly attenuates 5-CT-induced hypothermia in mice