Learn More
Medchemexpress LLC SB 258719 | 195199-95-2 | 338.51 | 50 MG
SDP
Catalog No. 5000445828
Encompass_Preferred
SB 258719 is a selective 5-HT7 receptor antagonist with a high affinity (pKi=7.5) for the receptor. It can be used for research in cancer and neurological diseases.
  • Selective 5-HT7 receptor antagonist
  • High affinity for the receptor (pKi=7.5)
  • Suitable for research in cancer
  • Suitable for research in neurological diseases
Catalog No. 50-004-45828 Supplier Medchemexpress LLC Supplier No. HYU0044350MG
SB 258719 is a selective 5-HT7 receptor antagonist with a high affinity (pKi=7.5) for the receptor. It can be used for research in cancer and neurological diseases.
  • Selective 5-HT7 receptor antagonist
  • High affinity for the receptor (pKi=7.5)
  • Suitable for research in cancer
  • Suitable for research in neurological diseases