Learn More
Medchemexpress LLC HY-U00443 5mg Medchemexpress, SB 258719 CAS:195199-95-2 Purity:>98%
SDP
Catalog No. 501934452
Encompass_Preferred
Medchemexpress, HY-U00443 5mg SB 258719 CAS:195199-95-2 SB 258719 is a selective 5-HT 7 receptor antagonist with a pK i of 7.5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4452 Supplier Medchemexpress LLC Supplier No. HYU004435MG
Medchemexpress, HY-U00443 5mg SB 258719 CAS:195199-95-2 SB 258719 is a selective 5-HT 7 receptor antagonist with a pK i of 7.5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.