Learn More
Medchemexpress LLC SB-265610 | 211096-49-0 | 98.98% | C14H9BrN6O | 50 MG
SDP
Catalog No. 5000359007
Encompass_Preferred
SB-265610 is a selective, competitive, nonpeptide, and allosteric CXCR2 antagonist. It blocks rat CINC-1-induced calcium mobilization (IC50 of 3.7 nM) and neutrophil chemotaxis (IC50 of 70 nM). In animal models, it significantly inhibits Gr-1+CD11b+ cell recruitment to mammary adenocarcinoma.
  • Selective, competitive, nonpeptide, and allosteric CXCR2 antagonist.
  • Blocks rat CINC-1-induced calcium mobilization (IC50 of 3.7 nM).
  • Blocks neutrophil chemotaxis (IC50 of 70 nM).
  • Reduces CINC-1's antiapoptotic effect.
  • Inhibits Gr-1+CD11b+ cell recruitment to mammary adenocarcinoma in animal models.
Catalog No. 50-003-59007 Supplier Medchemexpress LLC Supplier No. HY5068850MG
SB-265610 is a selective, competitive, nonpeptide, and allosteric CXCR2 antagonist. It blocks rat CINC-1-induced calcium mobilization (IC50 of 3.7 nM) and neutrophil chemotaxis (IC50 of 70 nM). In animal models, it significantly inhibits Gr-1+CD11b+ cell recruitment to mammary adenocarcinoma.
  • Selective, competitive, nonpeptide, and allosteric CXCR2 antagonist.
  • Blocks rat CINC-1-induced calcium mobilization (IC50 of 3.7 nM).
  • Blocks neutrophil chemotaxis (IC50 of 70 nM).
  • Reduces CINC-1's antiapoptotic effect.
  • Inhibits Gr-1+CD11b+ cell recruitment to mammary adenocarcinoma in animal models.