Learn More
American Radiolabeled Chemicals Inc SB 269970 3H 50UC
SDP
Catalog No. 50901245
Encompass_Preferred
SB 269970-3H hydrochloride 50uCi in ethanol, 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 388.95, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-245 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 182850UC
Add to Cart
Add to Cart
Request a Quote
SB 269970-3H hydrochloride 50uCi in ethanol, 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 388.95, Shipped in Dry Ice, Exclusive