Learn More
TARGETMOL CHEMICALS INC SB-269970 HYDROCHLORIDE 10MG
SDP
Catalog No. 502576480
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-269970 hydrochloride (SB-269970A) a hydrochloride salt form of SB-269970 is a 5-HT7 receptor antagonist (pKi of 8.3) and exhibits >50-fold selectivity against other receptors. purity: 98%
Catalog No. 50-257-6480 Supplier TARGETMOL CHEMICALS INC Supplier No. T665510MG
Also available in 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-269970 hydrochloride (SB-269970A) a hydrochloride salt form of SB-269970 is a 5-HT7 receptor antagonist (pKi of 8.3) and exhibits >50-fold selectivity against other receptors. purity: 98%