Learn More
Medchemexpress LLC HY-15370A 10mg Medchemexpress, SB-269970 (hydrochloride) CAS:261901-57-9 Purity:>98%
SDP
Catalog No. 502030616
Encompass_Preferred
Medchemexpress, HY-15370A 10mg SB-269970 (hydrochloride) CAS:261901-57-9 SB269970 hydrochloride (SB-269970A) is a hydrochloride salt form of SB-269970, which is a 5-HT7 receptor antagonist with the pKi of 8.3, exhibits >50-fold selectivity against other receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0616 Supplier Medchemexpress LLC Supplier No. HY15370A10MG
Medchemexpress, HY-15370A 10mg SB-269970 (hydrochloride) CAS:261901-57-9 SB269970 hydrochloride (SB-269970A) is a hydrochloride salt form of SB-269970, which is a 5-HT7 receptor antagonist with the pKi of 8.3, exhibits >50-fold selectivity against other receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.