Learn More
Medchemexpress LLC SB-269970 hydrochloride | 261901-57-9 | C18H29ClN2O3S | 5 MG
SDP
Catalog No. 5000376763
Encompass_Preferred
SB-269970 hydrochloride is a potent, selective, and brain-penetrant 5-HT7 receptor antagonist with a pKi of 8.3. It exhibits >50-fold selectivity against other 5-HT receptors.
  • Potent and selective 5-HT7 receptor antagonist
  • Brain-penetrant
  • Demonstrates greater than 50-fold selectivity against other 5-HT receptors
  • Significantly blocks amphetamine and ketamine-induced hyperactivity in animal models
Catalog No. 50-003-76763 Supplier Medchemexpress LLC Supplier No. HY15370A5MG
SB-269970 hydrochloride is a potent, selective, and brain-penetrant 5-HT7 receptor antagonist with a pKi of 8.3. It exhibits >50-fold selectivity against other 5-HT receptors.
  • Potent and selective 5-HT7 receptor antagonist
  • Brain-penetrant
  • Demonstrates greater than 50-fold selectivity against other 5-HT receptors
  • Significantly blocks amphetamine and ketamine-induced hyperactivity in animal models