Learn More
Medchemexpress LLC SB-269970 hydrochloride | 261901-57-9 | MFCD03788020 | C18H29ClN2O3S | 10 MM * 1 ML
SDP
Catalog No. 5000376102
Encompass_Preferred
SB-269970 hydrochloride is a potent, selective, and brain-penetrant antagonist of the 5-HT7 receptor. It demonstrates over 50-fold selectivity against other 5-HT receptors with a pKi of 8.3.
  • Potent and selective 5-HT7 receptor antagonist
  • Brain-penetrant
  • Exhibits over 50-fold selectivity against other 5-HT receptors
  • Significantly blocks amphetamine and ketamine-induced hyperactivity in animal models
Catalog No. 50-003-76102 Supplier Medchemexpress LLC Supplier No. HY15370A10MM/1ML
SB-269970 hydrochloride is a potent, selective, and brain-penetrant antagonist of the 5-HT7 receptor. It demonstrates over 50-fold selectivity against other 5-HT receptors with a pKi of 8.3.
  • Potent and selective 5-HT7 receptor antagonist
  • Brain-penetrant
  • Exhibits over 50-fold selectivity against other 5-HT receptors
  • Significantly blocks amphetamine and ketamine-induced hyperactivity in animal models