Learn More
Cayman Chemical SB-269970hydrochlorIde 50mg
SDP
Catalog No. 502967309
Encompass_Preferred
A potent 5-HT7A antagonist (pKi = 8.9) that demonstrates >50-fold binding selectivity over 5-HT5A and >250-fold selectivity over 5-HT1, 5-HT2, 5-HT4, 5-HT6, adrenergic α1, dopamine D2, and dopamine D3 receptors
Catalog No. 50-296-7309 Supplier Cayman Chemical Supplier No. 1708150 MG
A potent 5-HT7A antagonist (pKi = 8.9) that demonstrates >50-fold binding selectivity over 5-HT5A and >250-fold selectivity over 5-HT1, 5-HT2, 5-HT4, 5-HT6, adrenergic α1, dopamine D2, and dopamine D3 receptors