Learn More
Selleck Chemical LLC SB 271046 hydrochloride S2856-50mg
SDP
Catalog No. 503111860
Encompass_Preferred
SB 271046 hydrochloride is a potent selective and orally active 5-HT6 receptor antagonist with pKi of 8 9 exhibits 200-fold greater selectivity over other 5-HT receptor subtypes
Catalog No. 50-311-1860 Supplier Selleck Chemical LLC Supplier No. S285650MG
SB 271046 hydrochloride is a potent selective and orally active 5-HT6 receptor antagonist with pKi of 8 9 exhibits 200-fold greater selectivity over other 5-HT receptor subtypes