Learn More
Cayman Chemical SB-271046 (hydrochloride), Purity: ≥95%, CAS: 209481-24-3, 50 mg
SDP
Catalog No. 502968026
Encompass_Preferred
A selective, orally-available antagonist of the 5-HT6 receptor (pKi = 9.02-8.92); produces a significant increase in extracellular levels of glutamate and aspartate within the frontal cortex; used to evaluate the role of 5-HT6 in learning and memory
Catalog No. 50-296-8026 Supplier Cayman Chemical Supplier No. 1851350 MG
A selective, orally-available antagonist of the 5-HT6 receptor (pKi = 9.02-8.92); produces a significant increase in extracellular levels of glutamate and aspartate within the frontal cortex; used to evaluate the role of 5-HT6 in learning and memory