Learn More
Abcam SB-277011-A dihydrochloride, D3 antagonist, 25MG
Catalog No. 502707476
Encompass_Preferred
MW 511.5 Da, Purity >98%. Potent, highly selective D3 receptor antagonist (pKi = 7.95 at hD3 receptors). Exhibits over 100-fold selectivity over D2 receptors. Blood-brain barrier (BBB) permeable. Diminishes nicotine, cocaine and alcohol self-administration. Interferes with mechanisms of drug-taking and reinstatement of drug-taking behaviours.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7476 Supplier Abcam Supplier No. AB12056725MG
MW 511.5 Da, Purity >98%. Potent, highly selective D3 receptor antagonist (pKi = 7.95 at hD3 receptors). Exhibits over 100-fold selectivity over D2 receptors. Blood-brain barrier (BBB) permeable. Diminishes nicotine, cocaine and alcohol self-administration. Interferes with mechanisms of drug-taking and reinstatement of drug-taking behaviours.
The product is subject to the following: Abcam Restricted Use Statement