Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-277011 (hydrochloride) | 215804-67-4 | 99.59% | 475.02 | 10 MG
SDP

Catalog No. 5000346877
Encompass_Preferred

SB-277011 (hydrochloride) | 215804-67-4 | 99.59% | 475.02 | 10 MG

Catalog No. 50-003-46877 Supplier Medchemexpress LLC Supplier No. HY10847B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

SB-277011 (hydrochloride) | 215804-67-4 | 99.59% | 475.02 | 10 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.