Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-277011 (hydrochloride) | 215804-67-4 | 99.6% | 475.02 | 25 MG
SDP

Catalog No. 5000347663
Encompass_Preferred

SB-277011 hydrochloride is a potent, selective, orally bioavailable, and brain-penetrant dopamine D3 receptor (D3R) antagonist. It demonstrates 80- to 100-fold selectivity over other dopamine receptors.

  • Ki values of 10.7 nM and 11.2 nM at rodent and human D3R, respectively.
  • Selectivity over D2, 5-HT1B, and 5-HT1D receptors.
  • Oral bioavailability of 43% and a half-life of 2.0 h in rats.
  • High brain-penetrant (brain:blood ratio of 3.6:1).
  • Reverses quinelorane effects in the nucleus accumbens at 3 mg/kg (p.o.) in rats.
  • Reduces intravenous cocaine self-administration in rats (12.5-25 mg/kg, i.p.).
  • Inhibits basal and cocaine-enhanced locomotion in rats at 50 mg/kg.

Catalog No. 50-003-47663 Supplier Medchemexpress LLC Supplier No. 25MGHY10847B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB-277011 hydrochloride is a potent, selective, orally bioavailable, and brain-penetrant dopamine D3 receptor (D3R) antagonist. It demonstrates 80- to 100-fold selectivity over other dopamine receptors.

  • Ki values of 10.7 nM and 11.2 nM at rodent and human D3R, respectively.
  • Selectivity over D2, 5-HT1B, and 5-HT1D receptors.
  • Oral bioavailability of 43% and a half-life of 2.0 h in rats.
  • High brain-penetrant (brain:blood ratio of 3.6:1).
  • Reverses quinelorane effects in the nucleus accumbens at 3 mg/kg (p.o.) in rats.
  • Reduces intravenous cocaine self-administration in rats (12.5-25 mg/kg, i.p.).
  • Inhibits basal and cocaine-enhanced locomotion in rats at 50 mg/kg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.