Learn More
Medchemexpress LLC SB-277011 hydrochloride | 215804-67-4 | MFCD16875431 | 99.6% | 475.02 g·mol⁻¹ | C28H31ClN4O | 5 MG
SDP
Catalog No. 5000264725
Encompass_Preferred
SB-277011 hydrochloride is the hydrochloride salt of a selective dopamine D3 receptor antagonist, provided as a high-purity solid for preclinical pharmacology research. The product is suitable for formulation, in vitro, and in vivo studies, with recommended storage and handling conditions detailed in the supplier datasheet.
  • Selective dopamine D3 receptor antagonist activity
  • High purity (99.59% reported)
  • Solid powder form suitable for dosing or formulation
  • Reported oral bioavailability and brain penetration
  • Available in small research pack sizes including 5 mg
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month)
Catalog No. 50-002-64725 Supplier Medchemexpress LLC Supplier No. HY10847B5MG
Add to Cart
Add to Cart
SB-277011 hydrochloride is the hydrochloride salt of a selective dopamine D3 receptor antagonist, provided as a high-purity solid for preclinical pharmacology research. The product is suitable for formulation, in vitro, and in vivo studies, with recommended storage and handling conditions detailed in the supplier datasheet.
  • Selective dopamine D3 receptor antagonist activity
  • High purity (99.59% reported)
  • Solid powder form suitable for dosing or formulation
  • Reported oral bioavailability and brain penetration
  • Available in small research pack sizes including 5 mg
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month)