Learn More
Cayman Chemical SB 277011AhydrochlorIde 10mg
SDP
Catalog No. 502953932
Encompass_Preferred
A selective antagonist of the dopamine D3 receptor (pKi = 8.0); has high oral bioavailability and enters the central nervous system of the rat
Catalog No. 50-295-3932 Supplier Cayman Chemical Supplier No. 1198210 MG
A selective antagonist of the dopamine D3 receptor (pKi = 8.0); has high oral bioavailability and enters the central nervous system of the rat