Learn More
TARGETMOL CHEMICALS INC SB-366791 25MG
SDP
Catalog No. 502576398
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-366791 is a new and selective cinnamide TRPV1 antagonist. purity: 98%
Catalog No. 50-257-6398 Supplier TARGETMOL CHEMICALS INC Supplier No. T697725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-366791 is a new and selective cinnamide TRPV1 antagonist. purity: 98%