Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-399885 hydrochloride | 402713-81-9 | 98.3% | 482.81 g/mol | C18H22Cl3N3O4S | 1 ML
SDP

Catalog No. 5000246113
Encompass_Preferred

SB-399885 hydrochloride is a research reagent that acts as a selective 5-HT6 (serotonin 6) receptor antagonist. Supplied as the hydrochloride salt, it is suitable for in vitro and in vivo pharmacology studies and is available as solid material or as a ready-to-use 10 mM solution in DMSO.

  • Selective 5-HT6 receptor antagonist for research applications.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (98.3%).
  • Molecular weight 482.81 g/mol; molecular formula C18H22Cl3N3O4S.
  • CAS number 402713-81-9; supplied as the hydrochloride salt.

Catalog No. 50-002-46113 Supplier Medchemexpress LLC Supplier No. HY10309910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB-399885 hydrochloride is a research reagent that acts as a selective 5-HT6 (serotonin 6) receptor antagonist. Supplied as the hydrochloride salt, it is suitable for in vitro and in vivo pharmacology studies and is available as solid material or as a ready-to-use 10 mM solution in DMSO.

  • Selective 5-HT6 receptor antagonist for research applications.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (98.3%).
  • Molecular weight 482.81 g/mol; molecular formula C18H22Cl3N3O4S.
  • CAS number 402713-81-9; supplied as the hydrochloride salt.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.