Learn More
Medchemexpress LLC SB-399885 hydrochloride | 402713-81-9 | 98.3% | 482.81 g/mol | C18H22Cl3N3O4S | 10 MG
SDP
Catalog No. 5000247810
Encompass_Preferred
SB-399885 hydrochloride is a research-grade small molecule hydrochloride salt that acts as a selective 5-HT6 (serotonin) receptor antagonist. It is supplied as a solid for laboratory research, suitable for in vitro and in vivo pharmacology and behavioral studies. CAS 402713-81-9; molecular weight 482.81 g/mol.
  • High purity suitable for research use.
  • Hydrochloride salt form for improved solubility.
  • Applicable to in vitro receptor pharmacology and in vivo behavioral assays.
  • Supplied with a supplier datasheet for handling and storage information.
  • Available in small bench-scale pack sizes for preclinical studies.
Catalog No. 50-002-47810 Supplier Medchemexpress LLC Supplier No. HY10309910MG
SB-399885 hydrochloride is a research-grade small molecule hydrochloride salt that acts as a selective 5-HT6 (serotonin) receptor antagonist. It is supplied as a solid for laboratory research, suitable for in vitro and in vivo pharmacology and behavioral studies. CAS 402713-81-9; molecular weight 482.81 g/mol.
  • High purity suitable for research use.
  • Hydrochloride salt form for improved solubility.
  • Applicable to in vitro receptor pharmacology and in vivo behavioral assays.
  • Supplied with a supplier datasheet for handling and storage information.
  • Available in small bench-scale pack sizes for preclinical studies.