Learn More
Medchemexpress LLC SB-399885 hydrochloride | 402713-81-9 | 98.3% | 482.81 g/mol | C18H22Cl3N3O4S | 100 MG
SDP
Catalog No. 5000246076
Encompass_Preferred
SB-399885 hydrochloride is a research-grade small molecule that functions as a selective 5-HT6 receptor antagonist for laboratory pharmacology and neuroscience studies. It is supplied as a solid reagent and should be handled and stored according to standard laboratory safety practices.
  • Acts as a selective 5-HT6 receptor antagonist
  • Suitable for in vitro and in vivo pharmacology studies
  • Supplied as a solid with reported purity of 98.3%
  • Molecular weight 482.81 g/mol; CAS 402713-81-9
  • Available in multiple pack sizes for dose-ranging experiments
  • For research use only; not for human or clinical use
Catalog No. 50-002-46076 Supplier Medchemexpress LLC Supplier No. HY103099100MG
SB-399885 hydrochloride is a research-grade small molecule that functions as a selective 5-HT6 receptor antagonist for laboratory pharmacology and neuroscience studies. It is supplied as a solid reagent and should be handled and stored according to standard laboratory safety practices.
  • Acts as a selective 5-HT6 receptor antagonist
  • Suitable for in vitro and in vivo pharmacology studies
  • Supplied as a solid with reported purity of 98.3%
  • Molecular weight 482.81 g/mol; CAS 402713-81-9
  • Available in multiple pack sizes for dose-ranging experiments
  • For research use only; not for human or clinical use