Learn More
Medchemexpress LLC SB-399885 hydrochloride | 402713-81-9 | MFCD09970355 | 98.3% | 482.81 g·mol⁻¹ | C18H22Cl3N3O4S | 50 MG
SDP
Catalog No. 5000245797
Encompass_Preferred
SB-399885 hydrochloride is the hydrochloride salt of SB-399885, a selective 5-HT6 (serotonin 6) receptor antagonist supplied for research use. It is provided as a high-purity solid and is available as a ready-to-use solution in DMSO, for use in in vitro and in vivo pharmacological studies.
  • Selective 5-HT6 receptor antagonist.
  • High reported purity (~98.3%).
  • Hydrochloride salt form for improved stability and solubility.
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for in vitro and in vivo pharmacology studies.
Catalog No. 50-002-45797 Supplier Medchemexpress LLC Supplier No. HY10309950MG
SB-399885 hydrochloride is the hydrochloride salt of SB-399885, a selective 5-HT6 (serotonin 6) receptor antagonist supplied for research use. It is provided as a high-purity solid and is available as a ready-to-use solution in DMSO, for use in in vitro and in vivo pharmacological studies.
  • Selective 5-HT6 receptor antagonist.
  • High reported purity (~98.3%).
  • Hydrochloride salt form for improved stability and solubility.
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for in vitro and in vivo pharmacology studies.